Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Irgm2
Entrez Gene ID
54396
Gene Name
immunity-related GTPase family M member 2
Gene Aliases
AI481100, GTPI, Ifggd2, Iigp2
Chromosome Location
Chr.11: 58214977 - 58222783 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019440.3 NP_062313.3 1 - 2 193 72
GenBank mRNA AK142291.1 - 1 - 2 195 72
AK152870.1 - 1 - 2 134 72
AK153309.1 - 1 - 2 134 72
AK165347.1 - 1 - 2 131 72

View Full Product Details