Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sugp2
Entrez Gene ID
234373
Gene Name
SURP and G patch domain containing 2
Gene Aliases
Sfrs14, Srsf14, mKIAA0365
Chromosome Location
Chr.8: 70234209 - 70263106 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168290.1 NP_001161762.1 9 - 10 3528 100
NM_172755.3 NP_766343.3 9 - 10 3528 100
GenBank mRNA AF518875.1 - 9 - 10 3521 100
AK042293.1 - 9 - 10 3529 100
BC023276.1 - 5 - 6 1295 100
BC042763.1 - 7 - 8 1876 100
BC057305.1 - 9 - 10 3181 100

View Full Product Details