Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gabbr2
Entrez Gene ID
242425
Gene Name
gamma-aminobutyric acid (GABA) B receptor, 2
Gene Aliases
GABABR2, Gb2, Gm425, Gpr51
Chromosome Location
Chr.4: 46663898 - 46991714 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081141.1 NP_001074610.1 9 - 10 1660 73

View Full Product Details