Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Kcnd1
Entrez Gene ID
16506
Gene Name
potassium voltage-gated channel, Shal-related family, member 1
Gene Aliases
1110037K09Rik, Kca2-1, Kv4.1, Shal, Shal1, mShal1
Chromosome Location
Chr.X: 7823843 - 7836503 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK033805.1 - - 1034 69
BC139325.1 - 4 - 5 1549 69
BC139327.1 - 4 - 5 1549 69
M64226.1 - 4 - 5 1465 69

View Full Product Details