The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027324.5 | NP_081600.1 | 9 - 10 | 921 | 108 |
GenBank mRNA | AF325260.1 | - | 9 - 10 | 899 | 108 |
AK012650.1 | - | 9 - 10 | 920 | 109 | |
AK076205.1 | - | 4 - 5 | 290 | 108 | |
AK151110.1 | - | 9 - 10 | 867 | 108 | |
BC005743.1 | - | 9 - 10 | 878 | 108 |