Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Phf2
Entrez Gene ID
18676
Gene Name
PHD finger protein 2
Gene Aliases
GRC5
Chromosome Location
Chr.13: 48801750 - 48870885 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011078.3 NP_035208.2 11 - 12 1453 72
GenBank mRNA AB221591.1 - 11 - 12 1337 72
AF043726.1 - 11 - 12 1453 72
AK153906.1 - 11 - 12 1603 72
AK172995.1 - 9 - 10 1160 72
BC051633.1 - 11 - 12 1451 72

View Full Product Details