Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mfap2
Entrez Gene ID
17150
Gene Name
microfibrillar-associated protein 2
Gene Aliases
AI893631, Magp, Magp1
Chromosome Location
Chr.4: 141010424 - 141015984 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161799.1 NP_001155271.1 8 - 9 551 61
NM_008546.3 NP_032572.2 8 - 9 577 61
GenBank mRNA AK160213.1 - 8 - 9 553 61
AK164920.1 - 8 - 9 534 61
BC035490.1 - 8 - 9 511 61
DV654229.1 - 8 - 9 551 61
L23769.1 - 8 - 9 533 61

View Full Product Details