Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Trappc6b
Entrez Gene ID
78232
Gene Name
trafficking protein particle complex 6B
Gene Aliases
5830498C14Rik, C79212
Chromosome Location
Chr.12: 59043092 - 59061489 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK020026.1 - 1 - 2 286 76
AK046610.1 - 1 - 2 314 76
AK089083.1 - 1 - 2 127 76
BC031464.1 - 1 - 2 271 76

View Full Product Details