The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008831.4 | NP_032857.1 | 1 - 2 | 84 | 63 |
GenBank mRNA | AK152933.1 | - | 1 - 2 | 31 | 63 |
AK168248.1 | - | 1 - 2 | 32 | 63 | |
AK178178.1 | - | 1 - 2 | 38 | 63 |