Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vmn2r70
Entrez Gene ID
670940
Gene Name
vomeronasal 2, receptor 70
Gene Aliases
EG620835
Chromosome Location
Chr.7: 85558703 - 85569088 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105183.1 NP_001098653.1 3 - 4 1290 148

View Full Product Details