Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Rpl11
- Entrez Gene ID
-
67025
- Gene Name
- ribosomal protein L11
- Gene Aliases
-
2010203J19Rik
- Chromosome Location
-
Chr.4: 136049947 - 136053371 on Build GRCm38
- Gene Symbol
- Brf2
- Entrez Gene ID
-
66653
- Gene Name
- BRF2, subunit of RNA polymerase III transcription initiation factor, BRF1-like
- Gene Aliases
-
2700059M06Rik, 5730512K07Rik, AU018504, AU022602, BRFU, TFIIIB50
- Chromosome Location
-
Chr.8: 27123355 - 27128699 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC021918.1 |
- |
- |
1876 |
89 |
View Full Product Details