Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Agtr1b
Entrez Gene ID
11608
Gene Name
angiotensin II receptor, type 1b
Gene Aliases
AT1B, AT2R1B, Agtr-1b, Angtr-1b
Chromosome Location
Chr.3: 20314473 - 20367177 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175086.3 NP_780295.2 1 - 2 146 136
GenBank mRNA AK087228.1 - 1 - 2 146 136
BC109152.1 - 1 - 2 93 136
BC109153.1 - 1 - 2 93 136

View Full Product Details