Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cdk10
Entrez Gene ID
234854
Gene Name
cyclin-dependent kinase 10
Gene Aliases
BC017131
Chromosome Location
Chr.8: 123224840 - 123232256 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_194444.2 NP_919426.2 7 - 8 541 128
NM_194446.2 NP_919428.1 7 - 8 579 128
GenBank mRNA AK133918.1 - 7 - 8 534 128
AK144801.1 - 7 - 8 551 128
AK170594.1 - 7 - 8 575 128
BC017131.1 - 7 - 8 516 128
BC023736.1 - 7 - 8 560 128
BC052096.1 - 6 - 7 445 128
DQ288857.1 - 7 - 8 578 128

View Full Product Details