The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005525.2 | NP_001005525.1 | 6 - 7 | 845 | 102 |
GenBank mRNA | AK005213.1 | - | 4 - 5 | 576 | 102 |
BC132550.1 | - | 4 - 5 | 629 | 102 | |
BC132552.1 | - | 4 - 5 | 629 | 102 | |
BC145430.1 | - | 4 - 5 | 628 | 102 | |
DT902145.1 | - | 2 - 3 | 308 | 102 |