Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kif6
Entrez Gene ID
319991
Gene Name
kinesin family member 6
Gene Aliases
D130004B10Rik, D130084M03Rik
Chromosome Location
Chr.17: 49615104 - 49909847 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177052.3 NP_796026.2 21 - 22 2355 109
GenBank mRNA AK084082.1 - 6 - 7 2156 109

View Full Product Details