Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fau
Entrez Gene ID
14109
Gene Name
Finkel-Biskis-Reilly murine sarcoma virus (FBR-MuSV) ubiquitously expressed (fox derived)
Gene Aliases
MNSFbeta
Chromosome Location
Chr.19: 6057888 - 6059524 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190436.1 NP_001177365.1 1 - 2 360 159
GenBank mRNA AK008466.1 - 1 - 2 105 159

View Full Product Details