Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrrc8a
Entrez Gene ID
241296
Gene Name
leucine rich repeat containing 8A
Gene Aliases
Lrrc8, mKIAA1437
Chromosome Location
Chr.2: 30237769 - 30263790 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177725.4 NP_808393.1 3 - 4 2315 73

View Full Product Details