Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbxw14
Entrez Gene ID
50757
Gene Name
F-box and WD-40 domain protein 14
Gene Aliases
AU045528, E330009N23Rik, Fbx12, Fbxo12
Chromosome Location
Chr.9: 109270789 - 109287729 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015793.2 NP_056608.1 - 656 119
GenBank mRNA AK087709.1 - - 656 119
AK135682.1 - - 652 119
BC138018.1 - - 650 119

Gene Symbol
Fbxw28
Entrez Gene ID
668758
Gene Name
F-box and WD-40 domain protein 28
Gene Aliases
Gm9337
Chromosome Location
Chr.9: 109322886 - 109339659 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177419.1 NP_001170890.1 5 - 6 656 119
NM_001177420.1 NP_001170891.1 5 - 6 656 119
GenBank mRNA AK143322.1 - 5 - 6 652 119
AK162190.1 - 5 - 6 654 119
BC084592.1 - 5 - 6 647 119

View Full Product Details