The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003672.1 | NP_001003672.1 | 1 - 2 | 2563 | 98 |
GenBank mRNA | AY013769.1 | - | 1 - 2 | 2563 | 98 |