Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pnma3
Entrez Gene ID
245468
Gene Name
paraneoplastic antigen MA3
Gene Aliases
-
Chromosome Location
Chr.X: 73064787 - 73068191 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153169.2 NP_694809.1 1 - 1 3021 127
GenBank mRNA AK141560.1 - 1 - 1 2490 127
BC036726.1 - 1 - 1 3060 127

View Full Product Details