The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027128.2 | NP_081404.2 | 5 - 6 | 431 | 79 |
GenBank mRNA | AK009298.1 | - | 2 - 3 | 135 | 79 |
BC132394.1 | - | 2 - 3 | 75 | 79 | |
BC132396.1 | - | 2 - 3 | 74 | 79 | |
EF113909.1 | - | 5 - 6 | 431 | 79 |