Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Igsf9b
Entrez Gene ID
235086
Gene Name
immunoglobulin superfamily, member 9B
Gene Aliases
Gm508, mKIAA1030
Chromosome Location
Chr.9: 27299104 - 27357549 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033323.3 NP_001028495.2 13 - 14 2030 102
NM_001129787.1 NP_001123259.1 13 - 14 2030 102
GenBank mRNA AK158950.1 - 13 - 14 2030 102
AK220364.1 - 1 - 2 79 102
BC150882.1 - 13 - 14 1838 102
BC150887.1 - 13 - 14 1838 102

View Full Product Details