The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252569.1 | NP_001239498.1 | 6 - 6 | 1643 | 151 |
NM_009243.4 | NP_033269.1 | 5 - 5 | 1428 | 151 | |
GenBank mRNA | W82055.1 | - | 2 - 2 | 357 | 151 |