The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146542.2 | NP_666753.2 | 1 - 1 | 762 | 88 |
GenBank mRNA | BC111579.1 | - | 1 - 1 | 760 | 88 |