The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010930.1 | NP_001010930.1 | 2 - 3 | 199 | 84 |
GenBank mRNA | BC028645.1 | - | 1 - 2 | 141 | 84 |
BC087881.1 | - | 2 - 3 | 148 | 84 |