Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zbtb4
Entrez Gene ID
75580
Gene Name
zinc finger and BTB domain containing 4
Gene Aliases
2310026P19Rik, 9230111I22Rik, mKIAA1538
Chromosome Location
Chr.11: 69765912 - 69784026 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029348.2 NP_083624.2 3 - 3 853 121
GenBank mRNA AK147212.1 - 3 - 3 807 121
AK163593.1 - 3 - 3 916 121
AK173196.1 - 1 - 1 607 121
BC112421.1 - 2 - 2 649 121
BC113187.1 - 2 - 2 649 121

View Full Product Details