The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271725.1 | NP_001258654.1 | 12 - 12 | 7047 | 151 |
NM_001271726.1 | NP_001258655.1 | 12 - 12 | 7083 | 151 | |
NM_001271727.1 | NP_001258656.1 | 12 - 12 | 7352 | 151 | |
NM_001271728.1 | NP_001258657.1 | 12 - 12 | 7150 | 151 | |
NM_030706.3 | NP_109631.2 | 12 - 12 | 7146 | 151 | |
GenBank mRNA | AB043550.1 | - | 12 - 12 | 7127 | 151 |
AK054540.1 | - | 1 - 1 | 3552 | 151 | |
AK147275.1 | - | 12 - 12 | 7259 | 151 | |
AK147395.1 | - | 12 - 12 | 7141 | 151 | |
CA751006.1 | - | 1 - 1 | 674 | 151 |