The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166361.1 | NP_001159833.1 | 5 - 6 | 638 | 98 |
NM_001166362.1 | NP_001159834.1 | 4 - 5 | 551 | 98 | |
NM_001166363.1 | NP_001159835.1 | 4 - 5 | 518 | 98 | |
NM_010205.2 | NP_034335.1 | 4 - 5 | 710 | 98 | |
GenBank mRNA | AK131980.1 | - | 4 - 5 | 527 | 98 |
BC048734.1 | - | 5 - 6 | 628 | 98 | |
BU512456.1 | - | 4 - 5 | 586 | 98 | |
D12482.1 | - | 4 - 5 | 524 | 98 | |
D12483.1 | - | 4 - 5 | 709 | 98 | |
HQ260698.1 | - | 3 - 4 | 325 | 98 | |
U18673.1 | - | 4 - 5 | 402 | 98 | |
Z48746.1 | - | 4 - 5 | 378 | 98 |