Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hhla1
Entrez Gene ID
654498
Gene Name
HERV-H LTR-associating 1
Gene Aliases
F930104E18Rik
Chromosome Location
Chr.15: 65922443 - 65976804 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145096.1 NP_001138568.1 12 - 13 1031 60
GenBank mRNA AK158329.1 - 12 - 13 1033 60

View Full Product Details