Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mprip
Entrez Gene ID
26936
Gene Name
myosin phosphatase Rho interacting protein
Gene Aliases
9530046C02, AA536749, AI647711, C76423, RIP3, Rhoip3, mKIAA0864, p116Rip
Chromosome Location
Chr.11: 59662473 - 59780860 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012027.2 NP_036157.2 6 - 7 1074 58
NM_201245.3 NP_957697.1 6 - 7 1074 58
GenBank mRNA AK044512.1 - 6 - 7 824 58
AK154807.1 - 6 - 7 1081 58
AK155630.1 - 6 - 7 855 58
AK158296.1 - 6 - 7 1063 58
AK162530.1 - 3 - 4 405 58
AK170849.1 - 6 - 7 1058 58
AK171089.1 - 6 - 7 1063 58
BC039932.1 - 5 - 6 598 58
BC049803.1 - 6 - 7 600 58
BC063067.1 - 6 - 7 1037 58
U73200.1 - 6 - 7 934 58

View Full Product Details