Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Ncan
- Entrez Gene ID
-
13004
- Gene Name
- neurocan
- Gene Aliases
-
C230035B04, Cspg3, Cspg3-rs, Tgfbit
- Chromosome Location
-
Chr.8: 70093082 - 70120870 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK005742.1 |
- |
1 - 1 |
850 |
81 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 7066 |
Mir7066 |
Gm27290, mmu-mir-7066 |
-9.2kb
|
Anti-sense |
Chr. - 70102524 - 70102586 on Build GRCm38 |
Downstream |
regulatory factor X-associated ankyrin-containing protein |
Rfxank |
Tvl1 |
17.8kb
|
Anti-sense |
Chr. - 70130806 - 70139197 on Build GRCm38 |
View Full Product Details