Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Sri
- Entrez Gene ID
-
109552
- Gene Name
- sorcin
- Gene Aliases
-
2210417O06Rik, 2900070H08Rik, Sor
- Chromosome Location
-
Chr.5: 8046078 - 8069314 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK013762.1 |
- |
1 - 1 |
953 |
95 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CA876923.1 |
- |
1 |
|
|
Chr. 5 - 8062014 - 8062471 [-] on Build GRCm38 |
95 |
GenBankmRNA |
CJ289537.1 |
- |
1 |
166 |
|
Chr. 5 - 8062053 - 8062471 [+] on Build GRCm38 |
95 |
GenBankmRNA |
BY646206.1 |
- |
1 |
149 |
|
Chr. 5 - 8062070 - 8062471 [+] on Build GRCm38 |
95 |
GenBankmRNA |
AW538593.1 |
- |
1 |
|
|
Chr. 5 - 8061943 - 8062473 [-] on Build GRCm38 |
95 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
predicted gene 15731 |
Gm15731 |
OTTMUSG00000025798 |
-8.4kb
|
Anti-sense |
Chr. - 8037294 - 8052858 on Build GRCm38 |
Downstream |
a disintegrin and metallopeptidase domain 22 |
Adam22 |
2900022I03Rik, AI854032, MDC2 |
9.9kb
|
Anti-sense |
Chr. - 8072348 - 8368268 on Build GRCm38 |
View Full Product Details