Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Mip
- Entrez Gene ID
-
17339
- Gene Name
- major intrinsic protein of eye lens fiber
- Gene Aliases
-
Aqp0, Cat, Hfi, Lop, MIP26, Svl, shrivelled
- Chromosome Location
-
Chr.10: 128225838 - 128231811 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK140919.1 |
- |
- |
43 |
80 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BU525380.1 |
- |
|
98 |
|
Chr. - - [-] on Build |
80 |
GenBankmRNA |
CJ060815.1 |
- |
1 - 2 |
78 |
|
Chr. 10 - 128232320 - 128232826 [-] on Build GRCm38 |
80 |
GenBankmRNA |
CJ062130.1 |
- |
1 - 2 |
43 |
|
Chr. 10 - 128232060 - 128232790 [-] on Build GRCm38 |
80 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
timeless circadian clock 1 |
Timeless |
C77407, Debt69, tim |
|
Anti-sense |
Chr. - 128232063 - 128252941 on Build GRCm38 |
Downstream |
apolipoprotein N |
Apon |
AI195275, D10Ucla2 |
21.3kb
|
Anti-sense |
Chr. - 128254131 - 128255901 on Build GRCm38 |
View Full Product Details