Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- AV051173
- Entrez Gene ID
-
100502959
- Gene Name
- expressed sequence AV051173
- Gene Aliases
-
-
- Chromosome Location
-
Chr.4: 116684962 - 116686022 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_040442.1 |
|
2 - 3 |
276 |
62 |
GenBank mRNA |
AK131819.1 |
- |
2 - 3 |
277 |
62 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AI508296.1 |
- |
2 - 3 |
219 |
|
Chr. 4 - 116685044 - 116685954 [-] on Build GRCm38 |
62 |
GenBankmRNA |
CJ042621.1 |
- |
2 - 3 |
277 |
|
Chr. 4 - 116684964 - 116686022 [-] on Build GRCm38 |
62 |
GenBankmRNA |
BU936033.1 |
- |
|
230 |
|
Chr. - - [-] on Build |
62 |
GenBankmRNA |
BU583753.1 |
- |
2 - 3 |
220 |
|
Chr. 4 - 116684996 - 116685966 [-] on Build GRCm38 |
62 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
aldo-keto reductase family 1, member A1 (aldehyde reductase) |
Akr1a1 |
2610201A18Rik, Akr1a4 |
-33.3kb
|
Sense |
Chr. - 116636510 - 116651674 on Build GRCm38 |
Downstream |
methylmalonic aciduria cblC type, with homocystinuria |
Mmachc |
1810037K07Rik |
16.6kb
|
Sense |
Chr. - 116702434 - 116708385 on Build GRCm38 |
View Full Product Details