Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK156628.1 - 6052 Chr. - - [-] on Build 110
GenBankmRNA BY566700.1 - 1 254 Chr. X - 151825779 - 151826238 [+] on Build GRCm38 110
GenBankmRNA AU258485.2 - 1 321 Chr. X - 151825713 - 151826119 [+] on Build GRCm38 110
GenBankmRNA BY765700.1 - 1 522 Chr. X - 151825508 - 151826238 [+] on Build GRCm38 110


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream HECT, UBA and WWE domain containing 1 Huwe1 5430439H10Rik, AU041296, Arf-bp1, C430014N20Rik, C80292, Gm1718, Ib772, LASU1, Mule, Ureb1 Sense Chr. - 151800807 - 151935417 on Build GRCm38
Downstream microRNA let7f-2 Mirlet7f-2 Mirlet7f2, Mirnlet7f2, let-7f-2 86.2kb Sense Chr. - 151912346 - 151912428 on Build GRCm38

View Full Product Details