Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam160b2
Entrez Gene ID
239170
Gene Name
family with sequence similarity 160, member B2
Gene Aliases
G430067P06Rik, Rai16
Chromosome Location
Chr.14: 70583293 - 70599835 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_194345.1 NP_919326.1 2 - 3 175 99
GenBank mRNA BC050861.1 - 2 - 3 176 99
BU152183.1 - 2 - 3 175 99
CF170130.1 - 2 - 3 131 99

View Full Product Details