The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019915.2 | NP_064299.2 | 2 - 3 | 779 | 110 |
GenBank mRNA | AF016463.1 | - | 2 - 3 | 779 | 110 |
AF016465.1 | - | 2 - 3 | 779 | 110 | |
X87612.1 | - | 2 - 3 | 779 | 110 |