The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033380.3 | NP_001028552.1 | 1 - 1 | 1954 | 92 |
GenBank mRNA | AK134593.1 | - | 1 - 1 | 1902 | 92 |
AK137678.1 | - | 1 - 1 | 1956 | 92 | |
AK149729.1 | - | 1 - 1 | 1868 | 92 | |
BC150856.1 | - | 1 - 1 | 1605 | 92 | |
BC150864.1 | - | 1 - 1 | 1605 | 92 |