Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Vmn2r31
- Entrez Gene ID
-
100042591
- Gene Name
- vomeronasal 2, receptor 31
- Gene Aliases
-
-
- Chromosome Location
-
Chr.7: 7383985 - 7399627 on Build GRCm38
- Gene Symbol
- Vmn2r29
- Entrez Gene ID
-
76229
- Gene Name
- vomeronasal 2, receptor 29
- Gene Aliases
-
6430701C03Rik
- Chromosome Location
-
Chr.7: 7212996 - 7278289 on Build GRCm38
- Gene Symbol
- Vmn2r33
- Entrez Gene ID
-
624512
- Gene Name
- vomeronasal 2, receptor 33
- Gene Aliases
-
-
- Chromosome Location
-
Chr.7: 7550967 - 7566786 on Build GRCm38
- Gene Symbol
- Vmn2r38
- Entrez Gene ID
-
434110
- Gene Name
- vomeronasal 2, receptor 38
- Gene Aliases
-
-
- Chromosome Location
-
Chr.7: 9074796 - 9097765 on Build GRCm38
- Gene Symbol
- Vmn2r46
- Entrez Gene ID
-
100042894
- Gene Name
- vomeronasal 2, receptor 46
- Gene Aliases
-
-
- Chromosome Location
-
Chr.7: 9754636 - 9770571 on Build GRCm38
View Full Product Details