Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vmn2r52
Entrez Gene ID
384534
Gene Name
vomeronasal 2, receptor 52
Gene Aliases
EG384534
Chromosome Location
Chr.7: 10158652 - 10176286 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105191.1 NP_001098661.1 3 - 4 1303 94

View Full Product Details