Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vmn1r114
Entrez Gene ID
100042996
Gene Name
vomeronasal 1 receptor 114
Gene Aliases
ENSMUSG00000074323, Gm10669
Chromosome Location
Chr.7: 20811221 - 20812186 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166837.1 NP_001160309.1 1 - 1 155 101

Gene Symbol
Gm10666
Entrez Gene ID
100043037
Gene Name
predicted gene 10666
Gene Aliases
ENSMUSG00000074310, Vmn1r141
Chromosome Location
Chr.7: 22133776 - 22134741 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167573.1 NP_001161045.1 1 - 1 155 101

Gene Symbol
Gm4175
Entrez Gene ID
100043019
Gene Name
predicted gene 4175
Gene Aliases
Vmn1r133
Chromosome Location
Chr.7: 21760531 - 21761475 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167165.1 NP_001160637.1 1 - 1 155 101

Gene Symbol
Gm4214
Entrez Gene ID
100043079
Gene Name
predicted gene 4214
Gene Aliases
Vmn1r161
Chromosome Location
Chr.7: 22983729 - 22984673 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167164.1 NP_001160636.1 1 - 1 155 101

Gene Symbol
Vmn1r126
Entrez Gene ID
100043013
Gene Name
vomeronasal 1 receptor 126
Gene Aliases
Gm4170
Chromosome Location
Chr.7: 21300502 - 21301467 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166838.1 NP_001160310.1 1 - 1 155 101

Gene Symbol
Vmn1r93
Entrez Gene ID
404290
Gene Name
vomeronasal 1 receptor 93
Gene Aliases
V1rd21
Chromosome Location
Chr.7: 20138123 - 20139088 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207547.2 NP_997430.2 1 - 1 155 101

View Full Product Details