Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Serpinb6a
- Entrez Gene ID
-
20719
- Gene Name
- serine (or cysteine) peptidase inhibitor, clade B, member 6a
- Gene Aliases
-
4930482L21Rik, D330015H01Rik, PI-6, Serpinb6, Spi3, ovalbumin
- Chromosome Location
-
Chr.13: 33917918 - 34002794 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_009254.3 |
NP_033280.1 |
1 - 2 |
162 |
103 |
View Full Product Details