The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025939.2 | NP_080215.1 | 1 - 2 | 425 | 82 |
GenBank mRNA | AK012118.1 | - | 1 - 2 | 183 | 82 |
AK079282.1 | - | 1 - 2 | 84 | 82 | |
AK146722.1 | - | 2 - 3 | 377 | 82 | |
AK159398.1 | - | 1 - 2 | 230 | 82 | |
AK166264.1 | - | 1 - 2 | 80 | 82 | |
BC052691.1 | - | 1 - 2 | 425 | 82 |