Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pcdhb15
Entrez Gene ID
93886
Gene Name
protocadherin beta 15
Gene Aliases
Pcdhb7, PcdhbO
Chromosome Location
Chr.18: 37473546 - 37476340 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053140.3 NP_444370.1 1 - 1 2451 112
GenBank mRNA AK038663.1 - 1 - 1 2453 112
AY013776.1 - 1 - 1 2280 112
BC066178.1 - 1 - 1 2457 112

View Full Product Details