Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Padi6
Entrez Gene ID
242726
Gene Name
peptidyl arginine deiminase, type VI
Gene Aliases
Pad6, Padi5, ePAD
Chromosome Location
Chr.4: 140727355 - 140742643 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153106.2 NP_694746.2 13 - 14 1643 79
GenBank mRNA AF529423.1 - 13 - 14 1619 79
BC053724.1 - 13 - 14 1641 79

View Full Product Details