Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hist1h4d
Entrez Gene ID
319156
Gene Name
histone cluster 1, H4d
Gene Aliases
-
Chromosome Location
Chr.13: 23581602 - 23581969 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175654.2 NP_783585.1 1 - 1 151 112
GenBank mRNA BC119611.2 - 1 - 1 150 112

Gene Symbol
Hist1h4b
Entrez Gene ID
326620
Gene Name
histone cluster 1, H4b
Gene Aliases
-
Chromosome Location
Chr.13: 23756937 - 23757386 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC132186.1 - 1 - 1 235 112
BC132212.1 - 1 - 1 235 112

Gene Symbol
Hist1h4f
Entrez Gene ID
319157
Gene Name
histone cluster 1, H4f
Gene Aliases
-
Chromosome Location
Chr.13: 23551286 - 23551643 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC115446.1 - 1 - 1 144 112

Gene Symbol
Hist1h4i
Entrez Gene ID
319158
Gene Name
histone cluster 1, H4i
Gene Aliases
-
Chromosome Location
Chr.13: 22040960 - 22041362 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175656.3 NP_783587.1 1 - 1 190 112
GenBank mRNA AK011560.1 - 1 - 1 191 112
BC019757.1 - 1 - 1 180 112
BC052219.1 - 1 - 1 163 112
BC092144.1 - 1 - 1 180 112
BC125598.1 - 1 - 1 174 112
BC125600.1 - 1 - 1 174 112

Gene Symbol
Hist1h4m
Entrez Gene ID
100041230
Gene Name
histone cluster 1, H4m
Gene Aliases
Gm11275, OTTMUSG00000000446
Chromosome Location
Chr.13: 21811746 - 21812150 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195421.1 NP_001182350.1 1 - 1 189 112

Gene Symbol
Hist1h4n
Entrez Gene ID
319161
Gene Name
histone cluster 1, H4n
Gene Aliases
-
Chromosome Location
Chr.13: 21831793 - 21832158 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175657.2 NP_783588.1 1 - 1 151 112
GenBank mRNA BC115448.2 - 1 - 1 151 112
BC152397.1 - 1 - 1 151 112

View Full Product Details