Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hist1h2ae
Entrez Gene ID
319166
Gene Name
histone cluster 1, H2ae
Gene Aliases
1190022L06
Chromosome Location
Chr.13: 23570663 - 23571220 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK028026.1 - 1 - 1 551 200

View Full Product Details