Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrrtm4
Entrez Gene ID
243499
Gene Name
leucine rich repeat transmembrane neuronal 4
Gene Aliases
7530419J18Rik, A230052N11
Chromosome Location
Chr.6: 80018859 - 80810162 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178731.5 NP_848846.3 1 - 2 62 61
GenBank mRNA CJ059997.1 - 2 - 3 322 61

View Full Product Details