Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vmn2r75
Entrez Gene ID
546981
Gene Name
vomeronasal 2, receptor 75
Gene Aliases
EG546981
Chromosome Location
Chr.7: 86148042 - 86171724 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102578.1 NP_001096048.1 6 - 6 1953 103

View Full Product Details