Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rgs22
Entrez Gene ID
626596
Gene Name
regulator of G-protein signalling 22
Gene Aliases
-
Chromosome Location
Chr.15: 35960511 - 36140630 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195748.1 NP_001182677.1 2 - 3 96 63
GenBank mRNA BY092716.1 - 2 - 3 97 63

View Full Product Details